MagaDye™ 535 ddGTP
Supplier: AAT BIOQUEST
Certificates
About this item
Sanger sequencing, also known as the chain termination method, is a technique for DNA sequencing based upon the selective incorporation of chain-terminating dideoxynucleotides (ddNTPs) by DNA polymerase.
- In-house quality control
- Useful for sanger sequencing applications
Caution: For research use only.