Order Entry
Finland
ContactUsLinkComponent
ProLite™ His-Tag Protein Gel Staining Kit
ProLite™ His-Tag Protein Gel Staining Kit
Catalog # AATB18010
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
ProLite™ His-Tag Protein Gel Staining Kit
Catalog # AATB18010
Supplier:  AAT BIOQUEST
CAS Number:  
Specifications
  • Pack type:
    Plastic vial
  • Pk:
    10
  • Description:
    ProLite™ His-Tag Protein Gel Staining Kit "Green Fluorescence"
  • Pk Info:
    10 gels
Specifications
About this item
This kit requires very little hands-on time, allowing for rapid protein expression screening in a variety of gel types using an imaging equipment with a standard FITC filter.
  • Green fluorescence
  • Compatible with standard FITC filter
Kit provides a fast, sensitive, and highly specific fluorescent stain for visualizing His-tagged fusion proteins directly in a polyacrylamide gel following electrophoresis
It detects nanogram of His-tagged proteins directly in gels, thus eliminating the additional western blotting step.
Caution: For research use only.