Order Entry
Finland
ContactUsLinkComponent
Amplite® Fluorimetric Renin Assay Kit *Red Fluorescence*
Amplite® Fluorimetric Renin Assay Kit *Red Fluorescence*
Catalog # AATB13530
Supplier:  AAT Bioquest
CAS Number:  
undefined
Amplite® Fluorimetric Renin Assay Kit *Red Fluorescence*
Catalog # AATB13530
Supplier:  AAT Bioquest
CAS Number:  
Specifications
  • Pk:
    100 Tests
  • Description:
    Amplite™ fluorimetric renin assay kit *red fluorescence*
Specifications
About this item
Renin is an enzyme that participates in the body's renin-angiotensin system (RAS) that mediates extracellular volume and arterial vasoconstriction.
  • In-house QC testing
  • Sensitive
  • Suggested protocol available
Caution: For research use only.