

Acetic acid-[D₄] (99.5% D) for NMR spectroscopy
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1186-52-3
:
Formula:D₃CCOOD
MDL Number:MFCD00051051
:64.02 g/mol
Boiling Pt:115.5 °C (760 mmHg)
Melting Pt:15…16 °C
Density:1.13 g/cm³
Flash Pt:40 °C
Storage Temperature:Ambient
EINECS:214-693-4
UN:2789
ADR:8,II
:GHS05
:GHS02
Specification Test Results
- Appearance:Clear colorless Liquid
- Water:≤0.1 % (Coulometric)
- D-Enrichment:≥99.4 %