Order Entry
Finland
ContactUsLinkComponent
Tritirachium album proteinase K
Tritirachium album proteinase K
Supplier:  Merck
Certificates
Tritirachium album proteinase K
Supplier:  Merck
Certificates

Specifications

  • CAS No.:
    39450-01-6
  • Species:
    Tritirachium album
  • Storage conditions:
    Freezer
  • Enzyme name:
    Proteinase K

Specifications

Related Information