Order Entry
Finland
ContactUsLinkComponent
Woods metal
  765-0917
undefined
Woods metal
  765-0917
  • Description:
    Woods metal, 100 g
  • Pk:
    1
An alloy with an approximate composition of 48% by weight bismuth, 28% lead, 12% cadmium and 12% tin. The metal has an unusually low melting point (+65 ° C) and is used in the study of phase transformations, melting point determinations, etc.
  • Delivered in rod form