Order Entry
Finland
Orders LinkContactUsLinkComponent
Chrysoeriol

Chrysoeriol

Chrysoeriol
Certificates

2 Options Available Below

Product Details & Documents
Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • PROOTRCD444310-10G
  • PROOTRCD444310-1G
  • CAS No.:
  • 13988-26-6
  • 13988-26-6
  • Formula:
  • C₁₂H₁₂O₅
  • C₁₂H₁₂O₅
  • Shipping conditions:
  • RT
  • RT
  • Storage temperature:
  • 4 °C
  • 4 °C

Specifications

Product Family Options

Product Information

  • Molecular weight (Mw)
    Volume
    Pk
    Availability
    Price
  • PROOTRCD444310-10G
    Diethylene glycol cyclic phthalate
    Diethylene glycol cyclic phthalate
    236.22
    10 g
    10 g
    Log in for availability
    Log in for availability
    1 * 10 g
    1 * 10 g - 3 280,00 €
    3 280,00 €
    1 * 10 g
       3 280,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    PROOTRCD444310-10G
    CAS No.
    13988-26-6
    Formula
    C₁₂H₁₂O₅
    Shipping conditions
    RT
    Storage temperature
    4 °C
  • PROOTRCD444310-1G
    Diethylene glycol cyclic phthalate
    Diethylene glycol cyclic phthalate
    236.22
    1 g
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 453,00 €
    453,00 €
    1 * 1 g
       453,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    PROOTRCD444310-1G
    CAS No.
    13988-26-6
    Formula
    C₁₂H₁₂O₅
    Shipping conditions
    RT
    Storage temperature
    4 °C
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.