Order Entry
Finland
Orders LinkContactUsLinkComponent
Chrysoeriol

Chrysoeriol

Chrysoeriol (>90%), 300.26, 10 mg, 10 mg
Catalog # PROOTRCC432820-10
Chrysoeriol
Catalog # PROOTRCC432820-10
CAS Number:  
Chrysoeriol

Chrysoeriol (>90%), 300.26, 10 mg, 10 mg

CAS Number:  
Price
472,00 €
Log in for availability
1 * 10 mg
Product Details & Documents
Specifications

Specifications

  • Pk:
    10 mg
  • Description:
    Chrysoeriol (>90%)
  • Molecular weight (Mw):
    300.26
  • Volume:
    10 mg
  • Cat. No.:
    PROOTRCC432820-10
  • Formula:
    C₁₆H₁₂O₆
  • Storage conditions:
    Keep containers tightly closed in a dry, cool and well-ventilated place. Please refer to the manufacturer's certificate for specific storage and transport temperature conditions.
  • Physical form:
    Solid
  • Appearance:
    Solid
  • CAS No.:
    491-71-4
  • Colour:
    Yellow to dark yellow

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.