Order Entry
Finland
Orders LinkContactUsLinkComponent
ACE-2

ACE-2

Supplier:   AAT Bioquest
ACE-2
Supplier:  AAT Bioquest
Certificates
ACE-2

2 Options Available Below

Product Details & Documents

ACE2 (angiotensin-converting enzyme 2) is a metalloproteinase that requires a divalent cation positioned at the active site in order to perform catalysis.

  • High flexibility
  • Higher throughput
Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • AATB13555
  • AATB13556
  • Protein/peptide name:
  • ACE-2
  • ACE-2
  • Grade:
  • Molecular Biology Grade
  • Molecular Biology Grade
  • Molecular weight:
  • 696,7 g/mol
  • 696,7 g/mol
  • Molecular formula:
  • C₃₂H₃₆N₆O₁₂
  • C₃₂H₃₆N₆O₁₂
  • Tested applications:
  • Fluorescence
  • Fluorescence

Specifications

Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Plastic bottle
    1 mg
    Log in for availability
    Log in for availability
    1 * 1 mg
    1 * 1 mg - 299,00 €
    299,00 €
    1 * 1 mg
       299,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    AATB13555
    Protein/peptide name
    ACE-2
    Grade
    Molecular Biology Grade
    Molecular weight
    696,7 g/mol
    Molecular formula
    C₃₂H₃₆N₆O₁₂
    Tested applications
    Fluorescence
  • Plastic bottle
    10 mg
    Log in for availability
    Log in for availability
    1 * 10 mg
    1 * 10 mg - 748,00 €
    748,00 €
    1 * 10 mg
       748,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    AATB13556
    Protein/peptide name
    ACE-2
    Grade
    Molecular Biology Grade
    Molecular weight
    696,7 g/mol
    Molecular formula
    C₃₂H₃₆N₆O₁₂
    Tested applications
    Fluorescence