ReadiUse™ Preactivated HRP Maleimide
Catalog # AATB11026
Supplier: AAT BIOQUEST
CAS Number:
Specifications
- Pk:300 µG
- Description:ReadiUse™ preactivated HRP maleimide
Specifications
About this item
ReadiUse™ Preactivated maleimide is a thiol-reactive HRP derivative.
- Ready-to-use
- Highly useful antibody label
- Widely used for immunoassays
- Multiple applications
Caution: For research use only.