Horseradish Peroxidase *Optimized for Conjugation*
Catalog # AATB11021
Supplier: AAT BIOQUEST
CAS Number:
Specifications
- Pk:10 mg
- Description:Horseradish peroxidase *Optimized for conjugation*
Specifications
About this item
Horseradish peroxidase (HRP) is a glycoprotein with a molecular weight of ~44 Kd.
- Highly useful antibody label
- Widely used for immunoassays
- Multiple applications
- Highly purified
Caution: For research use only.