Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
OG488 maleimide
OG488 maleimide ≥90% [equivalent to Oregon Green® 488 maleimide]
OG488 maleimide ≥90% [equivalent to Oregon Green® 488 maleimide]
AATB715
:
AAT BIOQUEST
:
OG488 maleimide ≥90% [equivalent to Oregon Green® 488 maleimide]
AATB715
:
AAT BIOQUEST
:
Pk:
5 mg
Seq:
2210
:
:
463.35 g/mol