Order Entry
Finland
ContactUsLinkComponent
 
3,3'-Dioctadecyloxacarbocyanine perchlorate (DiO) ≥95%
  AATB22066
 :  
undefined
3,3'-Dioctadecyloxacarbocyanine perchlorate (DiO) ≥95%
  AATB22066
 :  

 

  • Pk:
    25 mg
  • Seq:
    2315

 

 

 :
 :881.721 g/mol
 :34215-57-1
 :C₅₃H₈₅ClN₂O₆