
MQAE (N-(Ethoxycarbonylmethyl)-6-methoxyquinolinium bromide) chloride indicator
Catalog # AATB21255
Supplier: AAT BIOQUEST
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:326.19 g/mol
ADR:5,II
:GHS07
CAS Number:162558-52-3
Formula:C₁₄H₁₆NO₃Br
MDL Number:MFCD00467854