Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Fura-8™ AM
Fura-8™ AM
Fura-8™ AM
Irritant
Fura-8™ AM
Supplier:
AAT BIOQUEST
Certificates
Expand image
Irritant
Fura-8™ AM
Supplier:
AAT BIOQUEST
About this item
:
:
951.9 g/mol
ADR:
5,II
:
GHS07