Order Entry
Finland
ContactUsLinkComponent
Cyanine 7 hydrazide ≥95% [equivalent to Cy7® hydrazide]
Cyanine 7 hydrazide ≥95% [equivalent to Cy7® hydrazide]
Catalog # AATB166
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
Cyanine 7 hydrazide ≥95% [equivalent to Cy7® hydrazide]
Catalog # AATB166
Supplier:  AAT BIOQUEST
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Seq:
    2070

Specifications

About this item

 :
 :924.92 g/mol