Order Entry
Finland
ContactUsLinkComponent
H-D-Ala-AMC·TFA ≥90%
Irritant
H-D-Ala-AMC·TFA ≥90%
Catalog # AATB13459
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
Irritant
H-D-Ala-AMC·TFA ≥90%
Catalog # AATB13459
Supplier:  AAT BIOQUEST
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    5 mg
  • Seq:
    2210

Specifications

About this item

 :
 :360.289 g/mol
ADR:5,II
 :GHS07
CAS Number:201847-52-1
Formula:C₁₅H₁₅F₃N₂O₅
MDL Number:MFCD00151971