Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-Methylumbelliferyl phosphate
4-Methylumbelliferyl phosphate ≥95%
4-Methylumbelliferyl phosphate ≥95%
Irritant
4-Methylumbelliferyl phosphate ≥95%
Supplier:
AAT BIOQUEST
Certificates
Expand image
Irritant
4-Methylumbelliferyl phosphate ≥95%
Supplier:
AAT BIOQUEST
About this item
CAS Number:
3368-04-5
:
Formula:
C₁₀H₉O₆P
MDL Number:
MFCD00016969
:
256.15 g/mol
ADR:
5,II
:
GHS07