Order Entry
Finland
ContactUsLinkComponent
 
 
Luminol (3-Aminophthalhydrazide) ≥95%
  AATB11050
 :  
undefined
 
Luminol (3-Aminophthalhydrazide) ≥95%
  AATB11050
 :  

 

  • Pack type:
    Plastic bottle
  • Pk:
    1 g
  • Seq:
    3047

 

 

 :
 :5,II
 :177.16 g/mol
 :GHS07
 :521-31-3
 :C₈H₇N₃O₂
 :MFCD00006890