
10-Acetyl-3,7-dihydroxyphenoxazine ≥95%
Catalog # AATB11000
Supplier: AAT BIOQUEST
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:25 mg
- Seq:2315
Specifications
About this item
:
:257.24 g/mol
ADR:5,II
:GHS07
CAS Number:119171-73-2
Formula:C₁₄H₁₁NO₄
MDL Number:MFCD00467861