Optics manual for TESS 1 and 2
765-1136
: PHYWE
- Description:Optics manual for TESS 1 + 2
- Pk:1
Describes 70 different experiments, on 288 pages, that can be performed in the areas.
- Mirrors
- Light refraction
- Lenses
- Colors
- The human eye
- Optical instruments
- Optical guard theory
- Spread of light
The components are delivered in a plywood box that fits in the drawer of the rolling table.