Order Entry
Finland
ContactUsLinkComponent
Sodium chloride, white, powder
Sodium chloride, white, powder
Catalog # YSIA069758
Supplier:  YSI ENVIRONMENTAL
CAS Number:  
Sodium chloride, white, powder
Catalog # YSIA069758
Supplier:  YSI ENVIRONMENTAL
CAS Number:  

Specifications

  • Pk:
    30 mL
  • Seq:
    2345

Specifications

About this item

 :
 :58.44 g/mol
Density:2.17
 :GHS07
CAS Number:7647-14-5
Formula:NaCl
MDL Number:MFCD00003477