Order Entry
Finland
ContactUsLinkComponent
SPINeasy® DNA Pro Kit for Soil, MP Biomedicals
SPINeasy® DNA Pro Kit for Soil, MP Biomedicals
Catalog # 116546050.
Supplier:  MP Biomedicals
CAS Number:  
undefined
SPINeasy® DNA Pro Kit for Soil, MP Biomedicals
Catalog # 116546050.
Supplier:  MP Biomedicals
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Method / Format:
    Enzymatic extraction
  • Sample type:
    Soil
  • Target molecule:
    DNA
  • Description:
    SPINeasy® DNA Pro kit for soil, 50 preps
  • Storage temperature:
    15...25 ℃

Specifications

About this item

Soil samples are complex environments characterised by the presence of inhibitory compounds, such as humic acid, heavy metals, and other aromatic components which may prove to be challenging for downstream analyses.

  • Effective isolation of high quality genomic DNA from high biomass and low biomass sample
  • Unbiased alpha diversity results
  • Higher purity and shorter processing time
  • Compatible with vacuum manifold

The SPINeasy® DNA Pro Kit for Soil has been carefully designed for the isolation of pure microbiome genomic DNA from challenging soil types including those with low biomass or those highly contaminated. The SPINeasy® DNA Pro Kit for Soil effectively lyses various microbiome population, including bacteria, fungi, viruses, and protists. The kit provides similar yields to that of our highly cited FastDNA™ SPIN Kit, but with improved purity and reduced processing time.

Isolated DNA products showed no inhibition in PCR and were immediately ready to be used in downstream applications, including long fragment PCR, qPCR, and next-generation sequencing (16S and whole genome) without the need of further inhibitor removal step.