Electrodefraction tubes
Electron diffraction tube
Catalog # 765-1204
Marketsource
Supplier: PHYWE
Jump to:
Product Details & Documents
Equipment for quantitative demonstration of the electron's wave nature and particle properties. At high electron energy, interference can be observed and evaluated in the form of Debye-Scherrer rings. The Broglie equation and Bragg's conditions can be proved. At low electron energies, the paticle properties of the electron can be demonstrated in a manner similar to geometric optics.
Cathode voltage: 6.3 V AC / 300 mA
Anode voltage: 2 V to 12 kV
Anode current: just over 1 mA
Specifications
- Description:Electron diffraction tube
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



