Order Entry
Finland
ContactUsLinkComponent
TD-NMR Glass Tubes
TD-NMR Glass Tubes
Catalog # MSPP1824512
TD-NMR Glass Tubes
Catalog # MSPP1824512

Specifications

  • Ext.Ø:
    18 mm
  • Description:
    TD-NMR Glass tubes
  • Length:
    180 mm
  • Seq:
    02
  • Pk:
    100

Specifications

About this item

TD-NMR Glass tubes with a lenght of 180 mm.

For 50 mm version, part number 1824514 has been replaced by a newer version with part number 1852653. Use and specs are identical.