DURAN® Stirring Rods, Glass
Supplier: Schott AG
Certificates
About this item
Very long glass rods made from Borosilicate 3.3 glass.
- High chemical resistance
- High hydrolytical resistance
- High temperature resistance
- High resistance to thermal shock
Very long glass rods made from Borosilicate 3.3 glass.