Order Entry
Finland
ContactUsLinkComponent
Influenza Virus B IgG (Human) ELISA kit
Influenza Virus B IgG (Human) ELISA kit
Catalog # ABNOKA6780.1KIT
Supplier:  Abnova
CAS Number:  
Influenza Virus B IgG (Human) ELISA kit
Catalog # ABNOKA6780.1KIT
Supplier:  Abnova
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Primary antibody reactivity:
    Human
  • Target protein:
    IgG
  • Assay Principle:
    Qualitative

Specifications

About this item

Influenza Virus B IgG (Human) ELISA Kit is a qualitative determination of IgG class antibodies against Influenza Virus B in human serum or plasma.