Order Entry
Finland
Orders LinkContactUsLinkComponent
Recombinant Human ALDOC (from E. coli)

Recombinant Human ALDOC (from E. coli)

20 µG
Catalog # ABNOP8034.20UG
Supplier:   Abnova
Recombinant Human ALDOC (from E. coli)
Catalog # ABNOP8034.20UG
Supplier:  Abnova
CAS Number:  
Recombinant Human ALDOC (from E. coli)

20 µG

CAS Number:  
Price
523,00 €
Log in for availability
1 * 1 Each
Product Details & Documents
Specifications

Specifications

  • Pk:
    20 µG
  • Cat. No.:
    ABNOP8034.20UG
  • Protein/peptide type:
    Recombinant
  • Protein/peptide name:
    ALDOC
  • Gene ID:
    P09972
  • Sequence:
    1a.a.-364a.a.
  • Species:
    Human
  • Source:
    E. coli

Specifications