Order Entry
Finland
ContactUsLinkComponent
Experimentation kits, protein fingerprinting
Experimentation kits, protein fingerprinting
Catalog # BE-408
Supplier:  G-Biosciences
undefined
Experimentation kits, protein fingerprinting
Catalog # BE-408
Supplier:  G-Biosciences
Specifications
  • Description:
    Protein Fingerprinting
  • Pk:
    1 KIT
Specifications
About this item
In this lab activity, students learn to perform simple protein isolation procedures to isolate the protein fingerprint from various fresh tissues. They will compare the protein fingerprints of four different tissues to understand that the function of a particular organ is due to the proteins that are localised to the specific organ. Also included in this kit are four dried protein samples (from mouse liver, brain, heart and lungs) to compare as a control, if fresh tissues cannot be obtained.
  • Manipulation of protein samples
  • Protein molecules as the building blocks of life
  • Protein electrophoresis for problem solving tasks
  • Protein diversity across the animal kingdom
Also required: Protein electrophoresis equipment.
Delivery: Supplied with components needed for hands-on experimentation for six workstations of 4 to 5 students or 24 to 30 students. Supplied with Teacher’s Guide and separate Student’s Guides.