Laser Optical Set - LOS1
LOS1, laser optical set
Catalog # 765-1011
Supplier: KVANT LIFE SCIENCE
Jump to:
Product Details & Documents
A set of optical and mechanical elements that are designed to demonstrate the principles of optics using a coherent laser light source. Students will be helped in their understanding of polarised light, diffraction and interference allowing them to reconstruct a holographic image.
- A magnetic table for secure positioning of the elements
- Plastic suitcase for secure transportation and storage
Delivery information: The set includes: diode laser 635nm - Pmax < 1mW (or 3mW); adjustable laser holder; 2 mirrors; 2 adjustable mirror holders; semitransparent mirror; 1 polarising filter; screen; ground screen; set of diffraction and interference structures; magnetic table; hologram; plastic suitcase; power supply: 110 to 240V/3V; battery box: 2x 1.5V AA battery type.
Specifications
- Description:LOS1, laser optical set