Data logger, Nova 5000 EX
Nova 5000 EX with built-in MultiLogPRO datalogger, MultiLab software, sensors and experiment guidance
Catalog # 765-0051
Supplier: FOURIER SYSTEMS
Jump to:
Product Details & Documents
Nova 5000 is a complete 1:1 PC solution featuring a Windows CE platform designed for school use. A small, full-featured student computer with data logger, word processing, spreadsheets, PowerPoint-type presentations, advanced graph plotting calculator, wireless internet connectivity. Battery can last for an entire working day. Weighing only 1,1 kg and measuring 24×19 cm, it fits both in the hands and in the backpack.
- Windows CE.NET 5,0 operating system with full Internet access, via Ethernet or built-in Wi-Fi 802,11b/g
- Supports web surfing and email
- Remote Desktop Connection for remote connection to a terminal server
- ActiveSync support, Windows Media Player
- Compatible with Fourier and Vernier sensors for data logging and analysis with MultiLab software
- Portrait four graphic calculator
- Administrative software features, including Explorer and Control Panel
- Word processing, spreadsheets and presentation support
- Handwriting recognition with Microsoft Transcriber
- Expandable memory with CompactFlash slot and USB ports
- Driver support for keyboard, mouse, printer, and more
- VGA connection to external monitor or projector
- Built-in speakers
Nova5000 is the missing link between notebook PCs and handheld PCs. The main focus is price, performance and user-friendliness. Nova 5000 can be used by anyone from simple measurements at primary school level to more advanced data processing at secondary school level.
Specifications
- Description:Nova 5000 EX with built-in MultiLogPRO datalogger, MultiLab software, sensors and experiment guidance
- Cat. No.:765-0051
- Interface:USB
Accessories
Product Information
- Catalog #AvailabilityPrice
- Carrying case for Nova5000
Product Information
- Catalog #AvailabilityPrice
- Rechargeable battery pack for Nova5000
Product Information
- Catalog #AvailabilityPrice
- USB communication cable, DT180