Dustless fingerprints, laboratory activity set
Product Details & Documents
Investigate alternate methods for lifting fingerprints.
- Note: The laboratory activity should be performed under a chemical fume hood
Students will gather data using fingerprint evidence as they observe, compare, contrast, and hypothesise their way through four different experimental procedures. They will learn how to make impressions in clay and pour a mold, lift fingerprints from a metal soda can using super glue fuming, and use both iodine and ninhydrin fuming to lift fingerprints from paper. The activity includes enough materials for six setups, an instructor’s manual, an assessment manual, and student data sheets.
Delivery information: The activity set includes enough materials for six groups, an instructor’s manual, an assessment manual, and student data sheets.
Specifications
- Description:Dustless fingerprints laboratory activity set