Analysis of drugs and poisons lab activity set
Catalog # 761-0297
Supplier: WARDS NATURAL SCIENCE ETABLISH
Specifications
- Description:Analysis of Drugs and Poisons, laboratory activity set
- Pk:1 KIT
Specifications
About this item
This set enables students to identify some common over-the-counter drugs, as well as learn how to test for controlled substances using simulated drugs. They will also analyse various metals and the amino acids found in simulated body fluids to detect heavy metal poisoning.
- Hands-on activity
- Multiple techniques
- Refill kit available to replenish consumables
An ultraviolet light source is required but not supplied in the set.
Delivery: Set includes materials for seven groups and an activity guide.