Order Entry
Finland
ContactUsLinkComponent
Human recombinant Wee1
Human recombinant Wee1
Catalog # 325896-10
CAS Number:  
Human recombinant Wee1
Catalog # 325896-10
CAS Number:  

Specifications

  • Pk:
    10 µG
  • Enzyme type:
    Recombinant
  • Species:
    Human
  • Enzyme name:
    Wee1

Specifications

About this item

Recombinant, human Wee1 fused at the N-terminus to a GST-His6-thrombin cleavage site sequence and expressed in S. frugiperda insect cells using a baculovirus expression system.

Wee1 is a tyrosine kinase that causes inhibitory phosporylation of Cdk1 and Cdk2 and acts as a mitotic inhibitor. Protects the nucleus from cytoplasmically activated cyclin B1-complexed CDC2 prior to the onset of mitosis.