About this item
The magnetic stirrer Hei-PLATE Mix 'n' Heat Expert convinces with its intuitive handling. You have an easily legible eye on all values and settings at any time.
The handling is made via two push-and-rotary controls with LED-rings for an easy identification of the active functions. Separate timers for heating and stirring functions create flexibility and let you control your experiments better. The user-friendly menu navigation makes working with this device particularly easy.
For controlling the temperature, you can choose between three heating modes: FAST for a particularly quick heating, as well as PRECISE and PRECISE+ for a particularly exact control of your target temperature without overshoot - this is how sensitive samples are optimally protected. Therefore, the device is compatible with the temperature sensor Pt1000 as well as the double sensor Pt1000 that allows the measurement in your sample and in the bath liquid simultaneously.
For the stirring speed, there is a three-stage setting (start-up behavior). Furthermore, you can set the rotational direction of the magnetic stirring bar and benefit from the automatic stir bar detector. As a result, your sample is optimally stirred at any time, even when there are irregularities while stirring.
This magnetic stirrer is equipped with the most important safety features. The large safety distance between the heating plate and the control panel, as well as the visual residual heat indicator at temperatures >50 °C, protect against burns.
Specifications
- Catalog No:
- HEID506-31100-00
- HEID506-32100-00
- Max. stirring capacity H₂O:
- 20 L
- 20 L
- Electrical:
- 230 V, 50/60 Hz
- 230 V, 50/60 Hz
- Speed display:
- 2×TFT display
- 2×TFT display
- Plate material:
- Kera-Disk®, ceramic coated Silumin (aluminium alloy)
- Kera-Disk®, ceramic coated Silumin (aluminium alloy)
- Plate dimensions:
- 145 mm
- 135 mm
- Weight:
- 3 kg
- 3 kg
- Overall dimensions:
- 168×299×106 mm
- 168×299×106 mm