Order Entry
Finland
ContactUsLinkComponent
96 Well Plate, RCH PSAXS
96 Well Plate, RCH PSAXS
  TCAT606-0501RCH
 :  Tecan
96 Well Plate, RCH PSAXS
  TCAT606-0501RCH
 :  Tecan
  • Description:
    Plate RCH PSAXS, 1 ml, 10 µm
  • Phase:
    PSAX-S
  • Filling:
    50 mg
  • Pk:
    1
Mixed mode reverse-phase with an anion exchanger for acidic compounds.
  • Ion-exchange
  • Liquid handler compatible
A 10 µm silica micro particulate with strong anionic exchange chemistry providing large contact zone for sorbent analyte interaction resulting in a fast and uniform sample prep.