Order Entry
Finland
ContactUsLinkComponent
Cerex® HPSAX 96 Well Plate RCH
Cerex® HPSAX 96 Well Plate RCH
Catalog # TCAT412-0401RCH
Supplier:  Tecan
Cerex® HPSAX 96 Well Plate RCH
Catalog # TCAT412-0401RCH
Supplier:  Tecan

Specifications

  • Description:
    Plate RCH Cerex HPSAX, 1 ml, 20 µm
  • Filling:
    40 mg
  • Pk:
    1

Specifications

About this item

Polymeric strong anionic exchange chemistry.

  • Ion-exchange
  • Liquid handler compatible

20 µm polymer micro particulate with strong anionic exchange chemistry providing large contact zone for sorbent analyte interaction resulting in fast and uniform sample prep. Ideal for acidic compounds.