Order Entry
Finland
ContactUsLinkComponent
Minigrip® Bags, Zip Seal, Topmatic
Minigrip® Bags, Zip Seal, Topmatic
Catalog # 129-3891
Supplier:  Flexico
Minigrip® Bags, Zip Seal, Topmatic
Catalog # 129-3891
Supplier:  Flexico

Specifications

  • Seq:
    10
  • Thickness:
    80 µm
  • Pk:
    250
  • W×L:
    360×250 mm

Specifications

About this item

It is made up of PE material with black closure.

  • Sensory effect and quality visual appearance

The plastic slider (POM) is generally not recyclable but it does not affect the bag's recyclability.