0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Finland
ContactUsLinkComponent
Furniture
Storage Cabinets
Accessories for Visible Storage Cabinet, 12 Drawers
Accessories for Visible Storage Cabinet, 12 Drawers
MULC14224V.04
:
MULTIROIR
Accessories for Visible Storage Cabinet, 12 Drawers
MULC14224V.04
:
MULTIROIR
Description:
Set of 10 transverse dividers for drawer, Outer dimensions: 92W×59H mm
Pk:
10