Order Entry
Finland
ContactUsLinkComponent
AseptiQuik® L Connectors
AseptiQuik® L Connectors
Supplier:  CPC COLDER
Certificates
undefined
AseptiQuik® L Connectors
Supplier:  CPC COLDER
Specifications
  • Catalog No:
  • CPCCAQL17012HT
  • CPCCAQL17012
  • CPCCAQL33024HT
  • CPCCAQL33024
  • CPCCAQL17016HT
  • 212-0823
  • Max. operating pressure:
  • Up to 4,1 bar; Up to 5,1 bar for 48 hours
  • Operating temperature:
  • 1...40 °C
Specifications
Related Information
Connector cuffs
Product Information