Cerex® WWP2 96 Well Plate, RCH
Catalog # TCAT417-0101RCH
Supplier: Tecan
Specifications
- Description:Plate RCH Cerex WWP2
- Phase:WWP2
- Pk:1
Specifications
About this item
Polymeric water-wettable resin ideal for reverse-phase retention of aqueous samples.
- Reversed-phase
- Liquid handler compatible
Special water wettable polymer with 300 Å pore size ideal for protein digest clean up and other protein related workflows.
Packaging: 1 piece per box