AseptiQuik® Sterile Connector Cuffs
Supplier: CPC COLDER
Certificates
About this item
AseptiQuik® connector cuffs provide additional support for more robust applications, so users can continue to transfer media easily with these quick and easy to use AseptiQuik® sterile connectors.
- Genderless design eases single-use system specifications with one-part number for both halves
- Flip-Click-Pull: intuitive three-step connection process reduces risk of operator error
- Audible confirmation of connection with no additional hardware required
- BPA-free
Use the AseptiQuik® cuffs to support use cases which have excessive sideload, pump, tension, vibration, or reduction in flow.
AseptiQuik® PPSU enables genderless connection for a greater range of chemical applications, offering versatile connections across downstream processes with a pH range from 2 to 12.