Order Entry
Finland
ContactUsLinkComponent
Banded Earplugs, X-fold
Banded Earplugs, X-fold
Catalog # UVEX2125.344
Supplier:  UVEX
Banded Earplugs, X-fold
Catalog # UVEX2125.344
Supplier:  UVEX

Specifications

  • Description:
    X-fold banded earplugs, SNR 23 dB
  • Pk:
    1

Specifications

About this item

The innovative design with unique, oval-shaped soft foam earplugs guarantees a comfortable fit.

  • X-fold is a lightweight banded ear protector weighing just 8 g
  • SNR 23 dB H: 26 dB, M: 18 dB, L: 17 dB
  • Colour: grey, sky blue, lime
  • Compact fold up, easy to carry
  • Suspended under the chin, eliminating friction sounds resulting from clothing
  • Band made up of polypropylene; earplugs made up of PU-foam
  • Thermal impac: Cold, heat

The foldable band is ergonomically shaped and prevents annoying noises from being transmitted by keeping the band away from the wearers clothes.