Zarate Trousers with ARC Protection
: SIOEN
Multifunctional and anti-static ARC trousers that provide excellent protection against arcs, heat, and metal splashes.
- Closure by 1 hidden button/front fly with zip closure
- 2 inset pockets with flap
- 1 back pocket with flap closed by concealed press studs
- 1 patched thigh pocket with flap closed by hidden press studs
- 1 ruler pocket with flap closed by concealed press studs
- Elasticated waist
- Knee pockets
- Waistband with belt loops
The trousers are made of a breathable and non-allergenic fabric and are very practical with two inset pockets with flap, one back pocket, one thigh pocket and one ruler pocket with concealed press stud fastening and reflective piping on the pocket flap. The trousers are provided with knee pockets where knee protection can be inserted.
Fabric: Sio-Safe™ Essential: 79% FR cotton+20% PES+1% AST; ±350 g/m²
: IEC 61482-2:2009 class 1- ATPV 15.2 cal/cm²; EN ISO 11612:2015/A1A2B1C1E3F1; EN ISO 11611:2015/Class 1 - A1 A2; EN 1149-5:2008; EN 13034:2005+A1:2009 Type PB [6]; EN ISO 13688:2013; Tested for harmful substances according to Oeko-Tex® Standard 100 (1708040/Centexbel).
- Catalog No:
- SIOE11VA2PFAM4456L
- SIOE11VA2PFAH4660R
- SIOE11VA2PFAM4456R
- SIOE11VA2PFAB9858S
- SIOE11VA2PFAH4664R
- SIOE11VA2PFAH4648S
- SIOE11VA2PFAH4650S
- SIOE11VA2PFAH4658S
- SIOE11VA2PFAM4448S
- SIOE11VA2PFAH4652S
- SIOE11VA2PFAH4654S
- SIOE11VA2PFAH4656S
- SIOE11VA2PFAH4654L
- SIOE11VA2PFAB9846L
- SIOE11VA2PFAB9860R
- SIOE11VA2PFAB9858R
- SIOE11VA2PFAB9864R
- SIOE11VA2PFAB9862R
- SIOE11VA2PFAM4458R
- SIOE11VA2PFAB9854L
- SIOE11VA2PFAH4644R
- SIOE11VA2PFAB9856L
- SIOE11VA2PFAH4662R
- SIOE11VA2PFAB9856S
- SIOE11VA2PFAB9852R
- SIOE11VA2PFAB9852L
- SIOE11VA2PFAB9848L
- SIOE11VA2PFAB9846R
- SIOE11VA2PFAB9850L
- SIOE11VA2PFAB9848R
- SIOE11VA2PFAH4654R
- SIOE11VA2PFAH4656L
- SIOE11VA2PFAB9844R
- SIOE11VA2PFAH4658R
- SIOE11VA2PFAB9850R
- SIOE11VA2PFAB9848S
- SIOE11VA2PFAB9850S
- SIOE11VA2PFAB9852S
- SIOE11VA2PFAB9854S
- SIOE11VA2PFAH4650R
- SIOE11VA2PFAM4454R
- SIOE11VA2PFAH4650L
- SIOE11VA2PFAB9854R
- SIOE11VA2PFAH4652L
- SIOE11VA2PFAH4646R
- SIOE11VA2PFAH4646L
- SIOE11VA2PFAH4648R
- SIOE11VA2PFAH4648L
- SIOE11VA2PFAM4450R
- SIOE11VA2PFAM4464R
- SIOE11VA2PFAM4452L
- SIOE11VA2PFAM4452R
- SIOE11VA2PFAM4454L
- SIOE11VA2PFAH4652R
- SIOE11VA2PFAM4446R
- SIOE11VA2PFAM4448L
- SIOE11VA2PFAH4656R
- SIOE11VA2PFAM4460R
- SIOE11VA2PFAM4448R
- SIOE11VA2PFAM4450L
- SIOE11VA2PFAM4462R
- SIOE11VA2PFAM4456S
- SIOE11VA2PFAM4458S
- SIOE11VA2PFAM4450S
- SIOE11VA2PFAM4452S
- SIOE11VA2PFAM4454S
- SIOE11VA2PFAM4444R
- SIOE11VA2PFAM4446L
- SIOE11VA2PFAB9856R
- Garment application:
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- Risk area:
- Anti-static
- Anti-static
- Anti-static
- Anti-static
- Anti-static
- Anti-static
- Anti-static
- Anti-static
- Anti-static
- Anti-static
- Anti-static
- Anti-static
- Anti-static
- Anti-static
- Anti-static
- Anti-static
- Anti-static
- Anti-static
- Anti-static
- Anti-static
- Anti-static
- Anti-static
- Anti-static
- Anti-static
- Anti-static
- Anti-static
- Anti-static
- Anti-static
- Anti-static
- Anti-static
- Anti-static
- Anti-static
- Anti-static
- Anti-static
- Anti-static
- Anti-static
- Anti-static
- Anti-static
- Anti-static
- Anti-static
- Anti-static
- Anti-static
- Anti-static
- Anti-static
- Anti-static
- Anti-static
- Anti-static
- Anti-static
- Anti-static
- Anti-static
- Anti-static
- Anti-static
- Anti-static
- Anti-static
- Anti-static
- Anti-static
- Anti-static
- Anti-static
- Anti-static
- Anti-static
- Anti-static
- Anti-static
- Anti-static
- Anti-static
- Anti-static
- Anti-static
- Anti-static
- Anti-static
- Anti-static
- Closure type:
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Fabric:
- Polyester/cotton
- Polyester/cotton
- Polyester/cotton
- Polyester/cotton
- Polyester/cotton
- Polyester/cotton
- Polyester/cotton
- Polyester/cotton
- Polyester/cotton
- Polyester/cotton
- Polyester/cotton
- Polyester/cotton
- Polyester/cotton
- Polyester/cotton
- Polyester/cotton
- Polyester/cotton
- Polyester/cotton
- Polyester/cotton
- Polyester/cotton
- Polyester/cotton
- Polyester/cotton
- Polyester/cotton
- Polyester/cotton
- Polyester/cotton
- Polyester/cotton
- Polyester/cotton
- Polyester/cotton
- Polyester/cotton
- Polyester/cotton
- Polyester/cotton
- Polyester/cotton
- Polyester/cotton
- Polyester/cotton
- Polyester/cotton
- Polyester/cotton
- Polyester/cotton
- Polyester/cotton
- Polyester/cotton
- Polyester/cotton
- Polyester/cotton
- Polyester/cotton
- Polyester/cotton
- Polyester/cotton
- Polyester/cotton
- Polyester/cotton
- Polyester/cotton
- Polyester/cotton
- Polyester/cotton
- Polyester/cotton
- Polyester/cotton
- Polyester/cotton
- Polyester/cotton
- Polyester/cotton
- Polyester/cotton
- Polyester/cotton
- Polyester/cotton
- Polyester/cotton
- Polyester/cotton
- Polyester/cotton
- Polyester/cotton
- Polyester/cotton
- Polyester/cotton
- Polyester/cotton
- Polyester/cotton
- Polyester/cotton
- Polyester/cotton
- Polyester/cotton
- Polyester/cotton
- Polyester/cotton