Clax Magic Multi 70C2 Stain Remover
Spot remover, 8,2, 6×0,75 L
DIVY100883039
: Diversey
Clax Magic Multi is a stain remover with excellent properties to remove a variety of stains such as make-up, lipstick, ink and curry stains. The product is specially formulated for use in commercial and on premise laundries and is ready to use on most types of fabric.
- Blend of solvents and other cleaning components
- Consists of four innovative stain-fighting solutions
- Kuvaus:Spot remover
- Pakkaus:6×0,75 L
- pH-arvo:8,2