Order Entry
Finland
ContactUsLinkComponent
Microflex® 92-134 Disposable Nitrile Examination Gloves
Microflex® 92-134 Disposable Nitrile Examination Gloves
Supplier:  Ansell
Certificates
Microflex® 92-134 Disposable Nitrile Examination Gloves
Supplier:  Ansell
Specifications
  • Catalog No:
  • ANSE92-134/M
  • ANSE92-134/S
  • ANSE92-134/XL
  • ANSE92-134/XS
  • ANSE92-134/XXL
  • ANSE92-134/L
  • Model:
  • Microflex® 92-134
  • Microflex® 92-134
  • Microflex® 92-134
  • Microflex® 92-134
  • Microflex® 92-134
  • Microflex® 92-134
  • Ambidextrous / hand-specific:
  • Ambidextrous
  • Ambidextrous
  • Ambidextrous
  • Ambidextrous
  • Ambidextrous
  • Ambidextrous
  • Disposable / reusable:
  • Disposable
  • Disposable
  • Disposable
  • Disposable
  • Disposable
  • Disposable
  • Latex / latex-free:
  • Latex-free
  • Latex-free
  • Latex-free
  • Latex-free
  • Latex-free
  • Latex-free
  • Powdered / powder-free:
  • Powder-free
  • Powder-free
  • Powder-free
  • Powder-free
  • Powder-free
  • Powder-free
  • Material:
  • Nitrile
  • Nitrile
  • Nitrile
  • Nitrile
  • Nitrile
  • Nitrile
  • Finish:
  • Textured fingers
  • Textured fingers
  • Textured fingers
  • Textured fingers
  • Textured fingers
  • Textured fingers
  • Cuff length:
  • Standard
  • Standard
  • Standard
  • Standard
  • Standard
  • Standard
  • Finger thickness:
  • 4.3 mil
  • 4.3 mil
  • 4.3 mil
  • 4.3 mil
  • 4.3 mil
  • 4.3 mil
  • Palm thickness:
  • 2.8 mil
  • 2.8 mil
  • 2.8 mil
  • 2.8 mil
  • 2.8 mil
  • 2.8 mil
  • FDA regulated:
  • +
  • +
  • +
  • +
  • +
  • +
  • Sterility:
  • Non sterile
  • Non sterile
  • Non sterile
  • Non sterile
  • Non sterile
  • Non sterile
  • Food contact approved:
  • +
  • +
  • +
  • +
  • +
  • +
  • ASTM No.:
  • D6319
  • D6319
  • D6319
  • D6319
  • D6319
  • D6319
Specifications