Order Entry
Finland
ContactUsLinkComponent
Chemist's Spoon
Chemist's Spoon
Catalog # USBE3406
undefined
Chemist's Spoon
Catalog # USBE3406
Specifications
  • Length:
    150 mm
  • Pk:
    1
Specifications
About this item
Single-ended chemical spoon made from stainless steel.
  • Autoclavable
  • Easy to clean
  • 16 mm width
  • 12 g weight
Excellent chemical resistance. High temperature resistance.