CS Cut F13 PU Gloves
: Portwest
Suitable for use with most touchscreen devices, this glove features a reinforced thumb crotch for extra durability.
- Level F cut resistance
- Protection against contact heat up to 100 °C
- Smooth PU coating for increased abrasion resistance
- Reflective label for improved visibility
- Seamless 13 gauge liner
- Palm dipped to increase dexterity and ventilation
- ANSI cut level A6
The breathable 13-gauge liner ensures an excellent fit and dexterity. PU Palm coating offers secure grip in dry and light oily handling environments. Reflective label increases glove visibility in low light conditions. Retail tag aids presentation for retail sales.
Colour: Black
: EN ISO 21420: 2020 Dexterity 5, EN 388: 2016 + A1: 2018 4X43F, EN 407: 2020 X1XXXX. ANSI/ISEA 105: 2016 CUT Level A6, ANSI/ISEA 105: 2016 ABRASION Level 6, ANSI/ISEA 105: 2016 PUNCTURE Level 3. CE certified.
- Catalog No:
- PRTWA670K8RXXL
- PRTWA670K8RXL
- PRTWA670K8RXS
- PRTWA670K8RS
- PRTWA670K8RL
- PRTWA670K8RM
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Latex / latex-free:
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Powdered / powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Material:
- HPPE/nylon/glass fibre/steel fibre
- HPPE/nylon/glass fibre/steel fibre
- HPPE/nylon/glass fibre/steel fibre
- HPPE/nylon/glass fibre/steel fibre
- HPPE/nylon/glass fibre/steel fibre
- HPPE/nylon/glass fibre/steel fibre
- Sterility:
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- Non sterile