Hi-Vis Flame Resistant Vests
Supplier: Portwest
Certificates
About this item
The FR75 is a great addition to our flame retardant vest collection.
- Premium sewn-on flame resistant reflective tape
- Hook and loop closure for easy access
- Non-magnetic; nickel- and ferrous-free
- Lightweight and comfortable
- Generous fit for wearer comfort
- CComplies with RIS 3279-TOM for rail industry (orange only)
Certified to EN ISO 20471 Class 2 and EN ISO 14116 Index 1, the vest must be worn over index 2 or 3 garments to offer flame retardant protection.
Hi-Vis FR75 vests are available in yellow or orange.
Certifications: EN ISO 14116 Index 1. EN ISO 20471 Class 2. RIS 3279 TOM Issue 2 (orange only). CE certified.
Specifications
- Catalog No:
- PRTWFR75YERL/XL
- PRTWFR75YER4X/5X
- PRTWFR75YERXX/3X
- PRTWFR75YERS/M
- Garment application:
- Fire protection
- Fire protection
- Fire protection
- Fire protection
- Risk area:
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Gender:
- Unisex
- Unisex
- Unisex
- Unisex
- Sleeve or leg length:
- Short
- Short
- Short
- Short
- Closure type:
- Hook-and-loop / VELCRO®
- Hook-and-loop / VELCRO®
- Hook-and-loop / VELCRO®
- Hook-and-loop / VELCRO®
- Fabric:
- Polyester
- Polyester
- Polyester
- Polyester
- Weight:
- 120 g/m²
- 120 g/m²
- 120 g/m²
- 120 g/m²