Thermo Scientific™ Matrix™ Sample Storage Starter Packs
Supplier: THERMO MATRIX TECHNOLOGIES
Certificates
About this item
Introduce automation into your workflow with Thermo Scientific™ sample storage starter packs.
- Eliminate the need to reformat non-2D cryotube samples
- Secure sample tracking with 2D, 1D, and human readable codes
- Scan 96 tubes in as little as 1 second with advanced and premium starter packs
- Help ensure quality with ISO 7 class cleanroom - manufactured tubes with sterility assurance
Choose from five different starter pack options.
Specifications
- Catalog No:
- 479-2466
- 479-2464
- 479-2467
- 479-2465
- 479-2468
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement